Ninhydrin 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

OR2351-25G OR2351-100G
76571-136EA 184.37 USD
76571-136 76571-132 76571-134
Ninhydrin 99%
Ninhydrin
Formula: C₉H₆O₄
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR