2-Pyrrolecarbaldehyde 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR23112-25G OR23112-100G
76570-910EA 321.36 USD
76570-910 76570-906 76570-908
2-Pyrrolecarbaldehyde 99%
2-Pyrrolecarbaldehyde
Formula: C₅H₅NO
Boiling Pt: 217…219 °C
Melting Pt: 43…46 °C
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005217
CAS Number: 1003-29-8
EINECS: 213-705-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR