4-Bromopyridinium chloride 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

OR23067-25G OR23067-500G OR23067-100G
76570-860EA 20.87 USD
76570-860 76570-864 76570-862
4-Bromopyridinium chloride 95+%
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
Melting Pt: 270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR