Tropine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

OR22908-100G OR22908-500G
76570-756EA 69.26 USD
76570-756 76570-758
Tropine
Tropine
Formula: C₈H₁₅NO
MW: 141.21 g/mol
Boiling Pt: 232…234 °C
Melting Pt: 61…65 °C
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00180667
CAS Number: 120-29-6
EINECS: 204-384-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR