6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR22802-25G OR22802-5G OR22802-1G
76570-742EA 231.24 USD
76570-742 76570-740 76570-738
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione 96%
6-Bromo-1,2,4-triazine-3,5(2H,4H)-dione
Formula: C₃H₂BrN₃O₂
MW: 191.98 g/mol
Melting Pt: 232…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00154757
CAS Number: 4956-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR