3-(N-Boc-Methylaminomethyl)azetidine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-[(Methylamino)methyl]azetidine hydrochloride, 3-BOC protected

OR2247-500MG OR2247-250MG
76570-650EA 255.66 USD
76570-650 76570-648
3-(N-Boc-Methylaminomethyl)azetidine hydrochloride
3-(N-Boc-Methylaminomethyl)azetidine hydrochloride
Formula: C₁₀H₂₁ClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09998721
CAS Number: 1170019-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR