3,5-Di-tert-butyl-4-hydroxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR21942-25G OR21942-100G
76570-452EA 20.34 USD
76570-452 76570-454
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 332
Melting Pt: 190
Storage Temperature: Ambient
MDL Number: MFCD00008826
CAS Number: 1620-98-0
EINECS: 216-592-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR