3,4,5-Tribromo-1H-pyrazole 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3,4,5-Tribromopyrazole
OR21421-100G
OR21421-25G
OR21421-5G
76570-236EA
114
USD
76570-236
76570-234
76570-232
3,4,5-Tribromo-1H-pyrazole 98%
3,4,5-Tribromo-1H-pyrazole
Formula:
C₃HBr₃N₂ MW: 304.78 g/mol Melting Pt: 184…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040248 CAS Number: 17635-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...