5-Chloro-2-nitropyridine 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR2032-25G OR2032-5G
76570-052EA 28.28 USD
76570-052 76570-050 76570-048
5-Chloro-2-nitropyridine 97+%
5-Chloro-2-nitropyridine
Formula: C₅H₃ClN₂O₂
Melting Pt: 120…123 °C
Storage Temperature: Ambient
MDL Number: MFCD03092916
CAS Number: 52092-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR