2'-Bromoacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2031-100G OR2031-25G OR2031-500G
76570-044EA 110.02 USD
76570-044 76570-042 76570-046
2'-Bromoacetophenone 98%
2'-Bromoacetophenone
Formula: C₈H₇BrO
Density: 1.476 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000067
CAS Number: 2142-69-0
EINECS: 218-398-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR