3-Methylxanthine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine

OR20005-250G OR20005-25G OR20005-100G
76569-932EA 206.78 USD
76569-932 76569-928 76569-930
3-Methylxanthine
3-Methylxanthine
Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
Melting Pt: 360 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005580
CAS Number: 1076-22-8
EINECS: 214-058-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR