4-Carboxy-3-chlorophenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Carboxy-3-chlorobenzeneboronic acid

OR1929-25G OR1929-5G OR1929-1G
76569-756EA 80.34 USD
76569-756 76569-754 76569-752
4-Carboxy-3-chlorophenylboronic acid 98%
4-Carboxy-3-chlorophenylboronic acid
Formula: C₇H₆BClO₄
MW: 200.39 g/mol
Melting Pt: 236…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656268
CAS Number: 136496-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR