4-(Diethylamino)benzoic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR18377-25G OR18377-5G OR18377-1G
76569-432EA 55.02 USD
76569-432 76569-430 76569-428
4-(Diethylamino)benzoic acid 99%
4-(Diethylamino)benzoic acid
Formula: C₁₁H₁₅NO₂
Melting Pt: 192…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00002538
CAS Number: 5429-28-7
EINECS: 226-582-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR