Ethylvanillin 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Ethoxy-4-hydroxybenzaldehyde, 3-(Ethoxy-d5)-4-hydroxybenzaldehyde

OR18302-500G OR18302-100G
76569-154EA 75.4 USD
76569-154 76569-152
Ethylvanillin 99%
Ethylvanillin
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 285
Melting Pt: 74…79 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR