6-Chlorooxindole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR18299-5G OR18299-25G
76569-146EA 41.2 USD
76569-146 76569-142 76569-144
6-Chlorooxindole 97%
6-Chlorooxindole
Formula: C₈H₆ClNO
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00209962
CAS Number: 56341-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR