5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR18276-100G OR18276-5G OR18276-25G
76569-082EA 110.02 USD
76569-082 76569-078 76569-080
5-Amino-1H-imidazole-4-carboxamide hydrochloride 98%
5-Amino-1H-imidazole-4-carboxamide hydrochloride
Formula: C₄H₇ClN₄O
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00012704
CAS Number: 72-40-2
EINECS: 200-778-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR