1,11-Undecanedioic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,9-Nonanedicarboxylic acid, 1,9-Nonanedicarboxylicacid, Nonamethylenedicarboxylic acid, Undecanedioic acid

OR16665-25G OR16665-100G
76568-258EA 20.34 USD
76568-258 76568-260
1,11-Undecanedioic acid
1,11-Undecanedioic acid
Formula: C₁₁H₂₀O₄
Melting Pt: 108…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00004444
CAS Number: 1852-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR