3-Amino-2,6-dimethoxypyridine 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR1585-5G OR1585-1G OR1585-250MG
76567-952EA 171.06 USD
76567-952 76567-950 76567-948
3-Amino-2,6-dimethoxypyridine 95+%
3-Amino-2,6-dimethoxypyridine
Formula: C7H10N2O2
Boiling Pt: 116…117 °C/10mmHg
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00832865
CAS Number: 28020-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR