4-Bromo-2-methoxythiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR15560-1G OR15560-5G
76567-814EA 75.4 USD
76567-814 76567-816
4-Bromo-2-methoxythiazole
4-Bromo-2-methoxythiazole
Formula: C₄H₄BrNOS
Storage Temperature: Refrigerator
MDL Number: MFCD07787374
CAS Number: 240816-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR