N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR1492-25G OR1492-10G OR1492-100G
76567-300EA 23.32 USD
76567-300 76567-298 76567-302
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Formula: C₁₈H₁₇NO₄
Melting Pt: 151…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062960
CAS Number: 79990-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR