1H-Pyrazole-1-carboximidamide hydrochloride
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine
OR14754-25G
OR14754-100G
76567-200EA
20.34
USD
76567-200
76567-202
1H-Pyrazole-1-carboximidamide hydrochloride
1H-Pyrazole-1-carboximidamide hydrochloride
Formula:
C₄H₇ClN₄ MW: 146.58 g/mol Melting Pt: 167…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210087 CAS Number: 4023-02-3 EINECS: 429-520-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...