Diethyl malonate 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR1354-500G
OR1354-10KG
OR1354-2.5KG
76566-388EA
20.32
USD
76566-388
76566-392
76566-390
Diethyl malonate 99%
Diethyl malonate
Formula:
C₇H₁₂O₄ Boiling Pt: 199 °C Melting Pt: –51 °C Density: 1.055 g/cm³ (25 °C) Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009195 CAS Number: 105-53-3 EINECS: 203-305-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...