2,2'-Bithiophene 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR1311-100G
OR1311-500G
OR1311-25G
76566-144EA
58.06
USD
76566-144
76566-146
76566-142
2,2'-Bithiophene 99%
2,2'-Bithiophene
Formula:
C₈H₆S₂ Boiling Pt: 260 °C Melting Pt: 32…33 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005414 CAS Number: 492-97-7 EINECS: 207-767-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...