N,O-Dimethylhydroxylamine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR12778-1KG OR12778-100G
76565-912EA 125.3 USD
76565-912 76565-910
N,O-Dimethylhydroxylamine hydrochloride 98%
N,O-Dimethylhydroxylamine hydrochloride
Formula: C₂H₈ClNO
MW: 97.55 g/mol
Melting Pt: 112…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00012485
CAS Number: 6638-79-5
EINECS: 229-642-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR