Homophthalic anhydride

Supplier: APOLLO SCIENTIFIC
Warning

OR12643-100G OR12643-25G OR12643-5G
76565-862EA 269.94 USD
76565-862 76565-860 76565-858
Homophthalic anhydride
Homophthalic anhydride
Formula: C₉H₆O₃
MW: 162.14 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR