3-Hydroxybenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR12009-500G OR12009-100G OR12009-1KG
76565-542EA 89.79 USD
76565-542 76565-540 76565-544
3-Hydroxybenzaldehyde 98%
3-Hydroxybenzaldehyde
Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 241
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003368
CAS Number: 100-83-4
EINECS: 202-892-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR