5-Hydroxy-1-indanone 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR11045-1G OR11045-10G OR11045-5G
76564-172EA 20.77 USD
76564-172 76564-176 76564-174
5-Hydroxy-1-indanone 97%
5-Hydroxy-1-indanone
Formula: C9H8O2
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00857527
CAS Number: 3470-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR