Potassium acetate 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-N-furfuryl-5-sulfamoylanthranlic acid, Diuretic salt, Acetic acid potassium salt, Acetic acid potassium salt

OR11032-1KG OR11032-10KG OR11032-5KG
76563-980EA 82.52 USD
76563-980 76563-984 76563-982
Potassium acetate 99%
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 303…307 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR