2-Phenylaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Aminobiphenyl, Biphenyl-2-ylamine, 2-Biphenylylamine

OR10963-5G OR10963-25G OR10963-100G
76563-552EA 42.6 USD
76563-552 76563-554 76563-556
2-Phenylaniline 98%
2-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 299 °C
Melting Pt: 47…50 °C
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR