4-Carboxyphenylboronic acid pinacol ester 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
OR10406-100G
OR10406-25G
OR10406-5G
76562-720EA
113.08
USD
76562-720
76559-538
76559-462
4-Carboxyphenylboronic acid pinacol ester 98%
4-Carboxyphenylboronic acid pinacol ester
Formula:
C₁₃H₁₇BO₄ Melting Pt: 227…231 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01863710 CAS Number: 180516-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...