4-(tert-Butoxycarbonyl)phenylboronic acid 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR10392-1G
OR10392-25G
OR10392-5G
76563-406EA
20.26
USD
76563-406
76561-702
76562-410
4-(tert-Butoxycarbonyl)phenylboronic acid 98%
4-(tert-Butoxycarbonyl)phenylboronic acid
Formula:
C₁₁H₁₅BO₄ Melting Pt: 100…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411946 CAS Number: 850568-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...