5-Chloroindole 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Chloro-1H-indole

OR10199-10G OR10199-25G OR10199-5G
76561-372EA 38.72 USD
76561-372 76561-458 76563-252
5-Chloroindole 97%
5-Chloroindole
Formula: C₈H₆ClN
MW: 151.6 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00005672
CAS Number: 17422-32-1
EINECS: 241-448-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR