4-Hydroxy-2-methoxybenzaldehyde 99.5+%

Supplier: APOLLO SCIENTIFIC
Warning

OR0857-1G OR0857-25G OR0857-5G
76562-426EA 20.32 USD
76562-426 76560-250 76560-898
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
4-Hydroxy-2-methoxybenzaldehyde
Formula: C₈H₈O₃
Melting Pt: 156…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00051964
CAS Number: 18278-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR