Malonic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Propanedioic acid

OR0786-100G OR0786-1KG OR0786-500G
76563-380EA 20.89 USD
76563-380 76561-110 76560-298
Malonic acid
Malonic acid
Formula: C₃H₄O₄
MW: 104.06 g/mol
Melting Pt: 132…136 °C (dec.)
Density: 1.62 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002707
CAS Number: 141-82-2
EINECS: 205-503-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR