5-(Phenylethynyl)-2-furoic acid, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR0707-1G OR0707-250MG
76560-292EA 131.8 USD
76560-292 76561-406 76561-976
5-(Phenylethynyl)-2-furoic acid, Technical Grade
5-(Phenylethynyl)-2-furoic acid
Formula: C₁₃H₈O₃
Melting Pt: 212…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00541949
CAS Number: 130423-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR