3-(2-Hydroxyethyl)indole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol

OR0519-100G OR0519-25G OR0519-5G
76562-250EA 600 USD
76562-250 76562-094 76562-254
3-(2-Hydroxyethyl)indole 98%
3-(2-Hydroxyethyl)indole
Formula: C₁₀H₁₁NO
Boiling Pt: 174 °C/2mmHg
Melting Pt: 57…60 °C
Flash Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00005659
CAS Number: 526-55-6
EINECS: 208-393-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR