2,7-Dichlorofluorene
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,7-Dichloro-9H-fluorene
OR0350-100G
OR0350-25G
OR0350-5G
76560-962EA
80.48
USD
76560-962
76561-290
76562-530
2,7-Dichlorofluorene
2,7-Dichlorofluorene
Formula:
C₁₃H₈Cl₂ Melting Pt: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00032840 CAS Number: 7012-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...