3-Acetylcoumarin

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Acetyl-2H-chromen-2-one

OR0090-1G OR0090-5G
76563-368EA 20.26 USD
76563-368 76559-708
3-Acetylcoumarin
3-Acetylcoumarin
Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00006853
CAS Number: 3949-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR