Potassium sorbate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

IN2901-1KG IN2901-5KG
76560-828EA 72.32 USD
76560-828 76561-764
Potassium sorbate 98%
Potassium sorbate
Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 210 °C (dec.)
Density: 1.36 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR