Deuterium oxide (99.9% D) for NMR spectroscopy

Supplier: VWR International

Synonyms: Water-D2, Heavy water

301401 301496 300368
BDH0906-25MLEA 77.03 USD
BDH0906-25ML BDH0906-10ML BDH0906-100ML
Deuterium oxide (99.9% D) for NMR spectroscopy
Deuterium oxide

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101 °C
Melting Pt: 3.81 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.90 %(mol)
1H-NMR Purity conform
TRIT max.74 Bq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR