Ohmeda Care Plus Incubator
Supplier: GUMBO MEDICAL
OCPI
470346-692EA 3272.69 USD
470346-692
Ohmeda Care Plus Incubator
Incubators Infant Incubators
Neonatal care.
  • Refurbished
  • 1 year parts warranty
  • For educational and training use only
The Care Plus® incubator delivers the latest technology for superior thermoregulation with an integrated microprocessor-based controller. This combination helps create the precise and consistent environment so important for the neonate while meeting all of the practical clinical needs of the health care provider including access to the infant.
Delivery information: This item has to ship by freight, Additional shipping charges may apply.
Caution: This product is refurbished and is intended for educational and training use only.