Cole-Parmer® BM-600 Planetary Mill High-Energy Ball Mill
Supplier: Avantor
76582-300EA 23425.82 USD
76582-300 76582-288 76582-302 76582-290 76582-292 76582-304 76582-306 76582-308 76582-310 76582-312 76582-294 76582-314 76582-316 76582-318 76582-320 76582-296 76582-298
Cole-Parmer® BM-600 Planetary Mill High-Energy Ball Mill
Mills
The Cole-Parmer® BM-600 planetary mill high-energy ball mill grinds samples with a mechanical motion rotating the jar in one direction while the platform, planetary wheel, often referred to as the sun wheel, is rotated in the opposite direction. The mechanical milling is performed at a 2:1 ratio causing the jar to rotate twice for every single rotation of the sun wheel. As the jar is moved, the relative centrifugal force is transferred to the grinding balls causing them to move in a circular motion, into each other and against the jar wall, grinding the sample. Functionally described as a high-shearing or a high-energy ball mill, this mill is used for pulverizing rocks, minerals, sand, cement, slag, ceramics, glass, and other brittle to hard samples to analytical fineness, blending powders or making emulsions.
The mills also feature a touch screen display, sleek modern design, forced-air cooling, a safety interlock, and a choice of hardened steel or stainless steel grinding jars and media, which are purchased separately.
Certifications: CE approved.