NoLimits DNA Fragments
Supplier: LIFE TECHNOLOGIES
SM1461 SM1481 SM1491 SM1621 SM1631 SM1641 SM1651 SM1671
470346-856EA 255.25 USD
470346-856 470346-866 470346-868 470346-858 470346-860 470346-854 470346-864 470346-862
NoLimits DNA Fragments
Educational Classroom Kits and Activities
Variable-sized DNA fragments for applications where a custom DNA size standard is required.
  • Broad range: 10 to 20000 bp
  • Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
  • Precise DNA concentration
  • Chromatography-purified individual DNA fragments
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology. Plasmid DNA, digested with Thermo Scientific Restriction Enzymes, is chromatography-purified to prepare the individual DNA fragments from the digestion mixture. The results are extremely pure DNA fragments, free of any truncated or degraded molecules. These custom DNA fragments are the best custom standards for applications ranging from electrophoresis (both gel and capillary) to HPLC and beyond.