1-(2-Bromoethoxy)-4-nitrobenzene 98%

Warning

38529-0050 38529-0250
TS38529-0050EA 27 USD
TS38529-0050 TS38529-0250
1-(2-Bromoethoxy)-4-nitrobenzene 98%
1-(2-Bromoethoxy)-4-nitrobenzene
Formula: C₈H₈BrNO₃
MW: 246.06
Melting Pt: 62…65 °C
MDL Number: MFCD00031363
CAS Number: 13288-06-7

Order Now

Specification Test Results

Appearance Yellow Powder or crystals
Infrared spectrum Conforms
Melting point 62 to 65 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR