4-Hydroxyphenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

36942-0010 36942-0050
TS36942-0010EA 55 USD
TS36942-0010 TS36942-0050
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS Number: 71597-85-8

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR