4-(Trifluoromethyl)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

35976-0050 35976-5000
TS35976-0050EA 157 USD
TS35976-0050 TS35976-5000
4-(Trifluoromethyl)phenylboronic acid 98%
4-(Trifluoromethyl)phenylboronic acid
Formula: C₇H₆BF₃O₂
Melting Pt: 245…250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151855
CAS Number: 128796-39-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR