6-Methoxy-m-toluidine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methoxy-5-methylaniline1-Amino-2-methoxy-5-methylbenzenep-Cresidine5-Methyl-o-anisidine2-Amino-4-methylanisole3-Amino-4-methoxytoluene

33208-1000 33208-5000 33208-0050 33208-0250
TS33208-5000EA 146.04 USD
TS33208-5000 TS33208-0050 TS33208-0250 TS33208-1000
6-Methoxy-m-toluidine 95%
6-Methoxy-m-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 235 °C (760 mmHg)
Melting Pt: 49…55 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00007815
CAS Number: 120-71-8
EINECS: 204-419-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Brown to black Crystalline chunks
Infrared spectrum Conforms
Melting point 49 to 55 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR