LunaScript RT SuperMix, New England Biolabs
Supplier: New England Biolabs (NEB)
M3010E M3010L M3010X
76559-404EA 7628 USD
76559-404 76559-406 76559-410
LunaScript RT SuperMix, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
LunaScript RT SuperMix is an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow. It features the thermostable Luna Reverse Transcriptase, which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is included to protect template RNA from degradation.
  • Contains random hexamer and oligo-dT primers, dNTPs, Murine RNase Inhibitor, and Luna RT
  • Less than 15 minute first-strand cDNA synthesis protocol
  • Featured in the ARTIC SARS-CoV-2 amplicon sequencing protocol from the ARTIC Network
  • Non-interfering, visible tracking dye helps to eliminate pipetting errors
  • Combine with Luna qPCR master mixes for robust RT-qPCR results
LunaScript RT SuperMix contains random hexamer and poly-dT primers, allowing even coverage across the length of the RNA targets. In addition, LunaScript RT SuperMix contains dNTPs and a blue tracking dye, providing a visual indicator that can be followed throughout the two-step RT-qPCR process. This product offers robust, linear, and sensitive detection using total RNA inputs as high as 1 µg and as low as single copies of RNA.