1-Boc-piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl Piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)piperidine, Piperidine-1-carboxylic acid tert-Butyl ester, tert-butyl piperidine-1-carboxylate, 1-Boc-piperidine
43808-0050
43808-0250
TS43808-0050EA
45
USD
TS43808-0050
TS43808-0250
1-Boc-piperidine 97%
1-Boc-piperidine
Formula:
C₁₀H₁₉NO₂ Boiling Pt: 58 °C (.3mbar) Flash Pt: 86 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093935 CAS Number: 75844-69-8 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...