6-Chloro-7-deazapurine 97%

Supplier: Thermo Scientific Chemicals
Danger

43744-0010 43744-0050
TS43744-0010EA 104 USD
TS43744-0010 TS43744-0050
6-Chloro-7-deazapurine 97%
6-Chloro-7-deazapurine
Formula: C₆H₄ClN₃
Melting Pt: 180.9…182.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01686865
CAS Number: 3680-69-1
UN: 2811

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR